missing translation for 'onlineSavingsMsg'
Learn More

Algestone Acetophenide, Mikromol™

Product Code. 17951216
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17951216 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17951216 Supplier Mikromol Supplier No. MM0476.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Algestone Acetophenide
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Algestone Acetophenide
CAS 24356-94-3
Analyt- eller komponentnavne Algestone Acetophenide
Forsendelsestilstand Room Temperature
Molekylær formel C29 H36 O4
Certificeringer/compliance ISO 17034
Synonym 2H-Naphth[1',2':6,7]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv., Algestone acetophenide, Alphasone acetophenide, Deladroxone, Dihydroxyprogesterone acetophenide, Droxone, Neolutin Depositum, P-DHP, ZK 9349, Pregn-4-ene-3,20-dione, 16,17-[[(1R)-1-phenylethylidene]bis(oxy)]-, (16alpha)-, Pregn-4-ene-3,20-dione, 16,17-[(1-phenylethylidene)bis(oxy)]-, [16alpha(R)]-, Pregn-4-ene-3,20-dione, 16alpha,17-dihydroxy-, cyclic acetal with acetophenone, (R)- (8CI)
InChI formel InChI=1S/C29H36O4/c1-18(30)29-25(32-28(4,33-29)19-8-6-5-7-9-19)17-24-22-11-10-20-16-21(31)12-14-26(20,2)23(22)13-15-27(24,29)3/h5-9,16,22-25H,10-15,17H2,1-4H3/t22-,23+,24+,25-,26+,27+,28-,29-/m1/s1
SMIL CC(=O)[C@@]12O[C@@](C)(O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@H]4CC[C@]23C)c6ccccc6
Molekylvægt (g/mol) 448.59
Mængde 250 mg
Formel vægt 448.2614 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.