missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ ALG5 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17142044 Shop alle Thermo Scientific produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17142044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17142044 Leverandør Invitrogen™ Leverandørnr. PA5114185

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: ALSYLEKRQK RDPAFTYEVI VVDDGSKDQT SKVAFKYCQK YGSDKVRVIT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.

Dolichyl-phosphate beta-glucosyltransferase (ALG5), belongs to the family of glycosyltransferases, specifically the hexosyltransferases. ALG5 is widely expressed in pancreas, placenta, liver, heart, brain, kidney, skeletal muscle, and lung. ALG5 can catalyze the chemical reaction between UDP-glucose and UDP with dolichyl phosphate.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ALG5
Applikationer Western Blot
Klassifikation Polyclonal
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering PBS with 2% sucrose and 0.09% sodium azide
Gene Alg5
Genadgangsnr. Q9Y673
Gene Alias 1500026A19Rik; 2600005J22Rik; ALG5; Alg5 homolog; ALG5, dolichyl-phosphate beta-glucosyltransferase; Alg5, S. cerevisiae, homolog of; asparagine-linked glycosylation 5 (dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5 homolog (S. cerevisiae, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5 homolog (yeast, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5, dolichyl-phosphate beta-glucosyltransferase homolog; asparagine-linked glycosylation protein 5 homolog; AV009521; AV170001; AW545564; bA421P11.2; dolichyl phosphate glucosyltransferase; dolichyl-phosphate beta-glucosyltransferase; DolP-glucosyltransferase; HSPC149
Gensymboler Alg5
Værtsarter Rabbit
Immunogen Immunogen sequence: ALSYLEKRQK RDPAFTYEVI VVDDGSKDQT SKVAFKYCQK YGSDKVRVIT (aa 87-136).
Oprensningsmetode Affinity Chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 29880
Målarter Human
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.