Få mere at vide
Alfuzosin-d3 Hydrochloride, TRC
Artikelnummer. 30322274
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30322274 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322274 Leverandør Toronto Research Chemicals Leverandørnr. TRCA53200210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alfuzosin-d3 Hydrochloride
CAS 1216383-97-9
Molekylær formel C19H25D3ClN5O4
Mængde 10 mg
Synonym N-[3-[(4-Amino-6,7-dimethoxy-2-quinazolynyl)(methyl-d3)amino]propyl]tetrahydro-2-furancarboxamide, Alfoten-d3, SL 77-499-10-d3, SL 77499-10-d3, Urion-d3, Uroxatral-d3, Xatral-d3
InChI formel InChI=1S/C19H27N5O4.ClH/c1-24(8-5-7-21-18(25)14-6-4-9-28-14)19-22-13-11-16(27-3)15(26-2)10-12(13)17(20)23-19;/h10-11,14H,4-9H2,1-3H3,(H,21,25)(H2,20,22,23);1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])N(CCCNC(=O)C1CCCO1)c2nc(N)c3cc(OC)c(OC)cc3n2
IUPAC navn N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-(trideuteriomethyl)amino]propyl]oxolane-2-carboxamide;hydrochloride
Molekylvægt (g/mol) 428.93