Få mere at vide
Alexidine-d10 Dihydrochloride, TRC
Artikelnummer. 30324639
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30324639 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324639 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5250021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alexidine-d10 Dihydrochloride
CAS 1246818-47-2
Molekylær formel C26H48D10Cl2N10
Mængde 1 mg
Synonym N,N''-Bis[2-(ethyl-d5)hexyl]-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide, 1,1'-Hexamethylene-bis[5-[2-(ethyl-d5)hexyl]biguanide], Bisguadine-d10, Bisguanidine-d10, Compound 904-d10, QR 711-d10, Sterwin 904-d10, Win 21904-d10
InChI formel InChI=1S/C26H56N10/c1-5-9-15-21(7-3)19-33-25(29)35-23(27)31-17-13-11-12-14-18-32-24(28)36-26(30)34-20-22(8-4)16-10-6-2/h21-22H,5-20H2,1-4H3,(H5,27,29,31,33,35)(H5,28,30,32,34,36)/i3D3,4D3,7D2,8D2
SMIL N=C(NC(NCC(C([2H])([2H])C([2H])([2H])[2H])CCCC)=N)NCCCCCCNC(NC(NCC(C([2H])([2H])C([2H])([2H])[2H])CCCC)=N)=N.Cl.Cl
IUPAC navn 1-[N'-[6-[[amino-[[N'-[2-(1,1,2,2,2-pentadeuterioethyl)hexyl]carbamimidoyl]amino]methylidene]amino]hexyl]carbamimidoyl]-2-[2-(1,1,2,2,2-pentadeuterioethyl)hexyl]guanidine
Molekylvægt (g/mol) 591.77