Få mere at vide
Alectinib M4 metabolite, TRC
Artikelnummer. 30134769
Skift visning
Mængde:
2.5 mg
5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30134769 5 mg -
30138548 10 mg -
30011566 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134769 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5211505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1256585-04-2
Molekylær formel C28H32N4O2
Molekylvægt (g/mol) 456.58
Synonym 9-Ethyl-6,11-dihydro-8-[4-[(2-hydroxyethyl)amino]-1-piperidinyl]-6,6-dimethyl-11-oxo-5H-benzo[b]carbazole-3-carbonitrile
IUPAC navn 9-ethyl-8-[4-(2-hydroxyethylamino)piperidin-1-yl]-6,6-dimethyl-11-oxo-5H-benzo[b]carbazole-3-carbonitrile
SMIL OCCNC1CCN(C2=CC(C(C)(C)C(C(C=CC(C#N)=C3)=C3N4)=C4C5=O)=C5C=C2CC)CC1
Tekniske data
Kemisk navn eller materiale Alectinib M4 metabolite
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C28H32N4O2/c1-4-18-14-21-22(15-24(18)32-10-7-19(8-11-32)30-9-12-33)28(2,3)27-25(26(21)34)20-6-5-17(16-29)13-23(20)31-27/h5-6,13-15,19,30-31,33H,4,7-12H2,1-3H3
Procent renhed >95%