Få mere at vide
Alectinib-d8, TRC
Artikelnummer. 30314150
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30314150 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314150 Leverandør Toronto Research Chemicals Leverandørnr. TRCA52118210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alectinib-d8
CAS 1256585-15-5
Renhedsgrad noter HPLC
Molekylær formel C30 D8 H26 N4 O2
Mængde 10 mg
Synonym 9-Ethyl-6,11-dihydro-6,6-dimethyl-8-[4-(4-morpholinyl-2,2,3,3,5,5,6,6-d8)-1-piperidinyl]-11-oxo-5H-benzo[b]carbazole-3-carbonitrile
InChI formel InChI=1S/C30H34N4O2/c1-4-20-16-23-24(17-26(20)34-9-7-21(8-10-34)33-11-13-36-14-12-33)30(2,3)29-27(28(23)35)22-6-5-19(18-31)15-25(22)32-29/h5-6,15-17,21,32H,4,7-14H2,1-3H3/i11D2,12D2,13D2,14D2
SMIL [2H]C1([2H])OC([2H])([2H])C([2H])([2H])N(C2CCN(CC2)c3cc4c(cc3CC)C(=O)c5c([nH]c6cc(ccc56)C#N)C4(C)C)C1([2H])[2H]
IUPAC navn 9-ethyl-6,6-dimethyl-8-[4-(2,2,3,3,5,5,6,6-octadeuteriomorpholin-4-yl)-1-piperidyl]-11-oxo-5H-benzo[b]carbazole-3-carbonitrile
Molekylvægt (g/mol) 490.66
Formel vægt 490.3184
Procent renhed >95