missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ ALDH Detection Reagent - BAAA-DA

Artikelnummer. 30040314
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30040314 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30040314 Leverandør Tocris Bioscience™ Leverandørnr. 7556/100U

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Fluorescent ALDH substrate and detection reagent. Used for: identification of cells with high ALDH activity. Application: flow cytometry, fluorescent-activated cell sorting (FACS)

Key information: ALDH Detection Reagent - BAAA-DA is a fluorescent substrate of aldehyde dehydrogenase (ALDH).

Used for: can be used in flow cytometry to sort cancer stem cells with high ALDH levels, and to isolate human hematopoietic progenitor cells. Can be used with efflux inhibitors such as Verapamil (Cat. No. 0654), Probenecid (Cat. No. 4107) or 2-deoxy-D-glucose (Cat. No. 4515) to block multi-drug resistance efflux.

Application: flow cytometry, fluorescent-activated cell sorting (FACS).

Properties and Photophysical Data: BAAA-DA is converted under acidic conditions to BAAA. BAAA, cell-permeant, is converted by intracellular ALDH into carboxylate ion amino acetate (BAA) and used to identify cells with high ALDH activity. Excitation and emission maxima (λ) are 505 nm and 512 nm, respectively; extinction coefficient = 74,595 M-1cm-1.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (T-4)-[N-(2,2-Diethoxyethyl)-5-[(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)methyl]-1H-pyrrole-2-propanamidato-κN1]difluoroboron
CAS 247069-93-8
Mængde 100 μg
Mål Enzyme Probes and Enzyme Substrates
Molekylær formel C20H28BF2N3O3
Renhed 0.95
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.