missing translation for 'onlineSavingsMsg'
Få mere at vide

Molecular Probes™ Aldehyde/Amidine Latex Beads, 4% w/v, 0.06μm

Artikelnummer. 11514077
Klik for at se tilgængelige muligheder
Mængde:
15 ml
Pakningsstørrelse:
15ml
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 11514077

Brand: Molecular Probes™ A37331

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Aldehyde/Amidine Latex Beads, 4% w/v, 0.06 μm

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

This aldehyde/amidine latex is similar to aldehyde/sulfate latex particles, but with a positively-charged amidine functional group to provide colloidal stability. This latex is not sensitive to multivalent cations.

Microspheres Are Available in Many Sizes and Surface Functionalities
  • 25 sizes to choose from—0.02 microns to 16 microns
  • 20 different surface—for ready conjugation to virtually any material
  • Surfactant free—ultraclean
  • Various quantities—from milliliter to 500-liter amounts

    Key Applications of Microspheres
  • Instrument calibration (flow cytometry, microscopy, HTS, HCS)
  • Flow testing (microfluidics, blood flow, water flow, and air flow)
  • Cell biology tracers (cell differentiation and cell tracing)
  • Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

    Get More Information
    Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

    We’ll Make a Custom Microsphere Product for You
    If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.

    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY

Tekniske data

Koncentration 0.04w/v
Indhold og opbevaring 2°C to 8°C
Oversigt Aldehyde/Amidine Latex Beads, 15mL, 4% w/v Concentration, 0.06μm Particle Size, White, Suspension Form, De-ionized Water Medium, Super Active Latex Beads, Room Temperature, 2 to 8°C Storage Temperature, Ideal Materials for the Adsorption of Materia
Farve White
Til brug med (applikation) Instrument Calibration (Flow Cytometry, Microscopy, HTS, HCS), Flow Testing (Microfluidics, Blood Flow, Water Flow, and Air Flow), Cell Biology Tracers (Cell Differentiation and Cell Tracing), Immunoassays (Agglutination Tests, ELISA, Particle Capture, and Contrast Reagents)
Materiale Polystyrene
Format Suspension
Emballage 1 Each
Parkeringsplads 351 A2/C(NH)NH2 (amidine group), 400 A2/CHO (aldehyde group)
Partikeldensitet 1.055g/cm3 at 20°C
Mængde 15 ml
Brydningsindeks 1.591 at 20°C
Overflademodifikation Aldehyde-amidine
Volumen (metrisk) 15 mL
Diameter (metrisk) middelværdi 0.06 μm
Diameter (metrisk) 0.06 μm
Vis mere Vis mindre

For Research Use Only. Not for use in diagnostic procedures.

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.