Få mere at vide
Alclometasone 17,21-Dipropionate, TRC
Artikelnummer. 30132472
Skift visning
Mængde:
2.5 mg
25 mg
100 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30132472 100 mg -
30138623 25 mg -
30003844 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132472 Leverandør Toronto Research Chemicals Leverandørnr. TRCA514580100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66734-13-2
Molekylær formel C28 H37 Cl O7
Molekylvægt (g/mol) 521.04
Synonym (7α,11β,16α)-7-Chloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Aclovate, Alclometasone 17,21-dipropionate, Alclometasone dipropionate, Almeta, Delonal, Sch 22219, Vaderm
IUPAC navn [2-[(7R,8S,9S,10R,11S,13S,14S,16R,17R)-7-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMIL CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@H](C)C[C@H]2[C@@H]3[C@H](Cl)CC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale Alclometasone 17,21-dipropionate
Analyt- eller komponentnavne Alclometasone 17,21-dipropionate
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C28H37ClO7/c1-6-22(33)35-14-21(32)28(36-23(34)7-2)15(3)10-18-24-19(29)12-16-11-17(30)8-9-26(16,4)25(24)20(31)13-27(18,28)5/h8-9,11,15,18-20,24-25,31H,6-7,10,12-14H2,1-5H3/t15-,18+,19-,20+,24-,25+,26+,27+,28+/m1/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 100 mg
Formel vægt 520.2228