missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
Alcian Blue 8GX increases in selectivity when increasing amounts of magnesium chloride are added. In contrast, studies show that when electrolyte concentration is very low, it has trouble binding to carboxylate or phosphate groups.
- Quantitatively determine glycosaminoglycans by forming insoluble complexes in solution
- Determine proteins based on their Resonance Light Scattering by strongly enhancing the signals
- Useful as a bacterial stain and as a dye for histiocytes and fibroblasts
- Used as a heteroglycan stain for neutral, sulfated and phosphated mucopolysaccharides and glycosaminoglycans in tissues such as cartilage and extracellular matrices
Tekniske data
Tekniske data
| CAS | 75881-23-1 |
| Molekylær formel | C56H68Cl4CuN16S4 |
| Synonym | Alcian Blue, Ingrain Blue 1 |
| Opløselighedsinformation | Soluble in absolute ethanol, cellosolve, ethylene glycol, water (8mg/mL) or ethanol (1mg/mL); practically insoluble in xylene |
| InChI nøgle | CKLBXIYTBHXJEH-UHFFFAOYSA-J |
| SMIL | CN(C)C(=[N+](C)C)SCC1=CC2=C(C=C1)C3=NC4=NC(=NC5=C6C=C(C=CC6=C([N-]5)N=C7C8=C(C=CC(=C8)CSC(=[N+](C)C)N(C)C)C(=N7)N=C2[N-]3)CSC(=[N+](C)C)N(C)C)C9=C4C=C(C=C9)CSC(=[N+](C)C)N(C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Cu+2] |
| Molekylvægt (g/mol) | 1298.858 |
| PubChem CID | 16211091 |
| Formel vægt | 1298.88 |
| Mængde | 25 g |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?