Få mere at vide
Beskrivelse
Alcian Blue 8Gx can be used to determine proteins based on their Resonance Light Scattering by strongly enhancing the signals. It is useful as a bacterial stain, and as a dye for histiocytes and fibroblasts. This product is used as a heteroglycan stain for neutral, sulfated and phosphated mucopolysaccharides and glycosaminoglycans in tissues such as cartilage and extracellular matrices. Studies show that when electrolyte concentration is very low, Alcian Blue 8Gx has trouble binding to carboxylate or phosphate groups.
Tekniske data
Tekniske data
| CAS | 75881-23-1 |
| Molekylær formel | C56H68Cl4CuN16S4 |
| Synonym | Alcian Blue, Ingrain Blue 1 |
| Opløselighedsinformation | Soluble in absolute ethanol, cellosolve, ethylene glycol, water (8mg/mL) or ethanol (1mg/mL); practically insoluble in xylene |
| InChI nøgle | CKLBXIYTBHXJEH-UHFFFAOYSA-J |
| SMIL | CN(C)C(=[N+](C)C)SCC1=CC2=C(C=C1)C3=NC4=NC(=NC5=C6C=C(C=CC6=C([N-]5)N=C7C8=C(C=CC(=C8)CSC(=[N+](C)C)N(C)C)C(=N7)N=C2[N-]3)CSC(=[N+](C)C)N(C)C)C9=C4C=C(C=C9)CSC(=[N+](C)C)N(C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Cu+2] |
| Molekylvægt (g/mol) | 1298.858 |
| PubChem CID | 16211091 |
| Formel vægt | 1298.88 |
| Mængde | 5 g |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.