missing translation for 'onlineSavingsMsg'
Få mere at vide

Albendazole Impurity G, TRC

Artikelnummer. 30466927
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30466927 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466927 Supplier Toronto Research Chemicals Supplier No. TRCM2887352.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl (5-Chloro-1H-benzimidazol-2-yl)carbamate
CAS 20367-38-8
Molekylær formel C9 H8 Cl N3 O2
Mængde 2.5 mg
Synonym N-(6-chloro-1H-benzimidazol-2-yl)-carbamic acid methyl ester, 2-Benzimidazolecarbamic acid, 5-chloro-, methyl ester (8CI), Carbamic acid, (5-chloro-1H-benzimidazol-2-yl)-, methyl ester (9CI), 2-[(Methoxycarbonyl)amino]-5-chlorobenzimidazole, G 981, Methyl (5-chloro-2-benzimidazolyl)carbamate
InChI formel InChI=1S/C9H8ClN3O2/c1-15-9(14)13-8-11-6-3-2-5(10)4-7(6)12-8/h2-4H,1H3,(H2,11,12,13,14)
SMIL COC(=O)Nc1nc2cc(Cl)ccc2[nH]1
IUPAC navn methyl N-(5-chloro-1H-benzimidazol-2-yl)carbamate
Molekylvægt (g/mol) 225.63
Formel vægt 225.0305
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.