Få mere at vide
Albendazole-d7, TRC
Artikelnummer. 30318557
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30318557 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318557 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5116122.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Albendazole-d7
CAS 1287076-43-0
Molekylær formel C12H8D7N3O2S
Mængde 2.5 mg
Synonym N-[6-(Propylthio-d7)-1H-benzimidazol-2-yl]carbamic Acid Methyl Ester, [5-(Propylthio-d7)-1H-benzimidazol-2-yl]carbamic Acid Methyl Ester, Albenza-d7, Andazol-d7, Ashialben-d7, Atasol-d7, Bruzol-d7, Eskazole-d7, Loveral-d7, Lurdex-d7, Proftril-d7, Valbazen-d7, Zentel-d7, NSC 220008-d7, Tobend-d7 Valbazen-d7, Vermisen-d7, Vermitan-d7, Vermizole-d7, Zentel-d7
InChI formel InChI=1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)/i1D3,3D2,6D2
SMIL O=C(OC)NC1=NC2=CC(SC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=CC=C2N1
IUPAC navn methyl N-[6-(1,1,2,2,3,3,3-heptadeuteriopropylsulfanyl)-1H-benzimidazol-2-yl]carbamate
Molekylvægt (g/mol) 272.37
Formel vægt 272.1324