Få mere at vide
Albendazole-d3 (Methoxy-d3), TRC
Artikelnummer. 30323542
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30323542 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323542 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5116132.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Albendazole methoxy-d3
CAS 1353867-92-1
Molekylær formel C12 H15 N3 O2 S
Mængde 2.5 mg
Synonym Trideuteriomethyl N-(5-propylsulfanyl-1H-benzimidazol-2-yl)carbamate
InChI formel InChI=1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)/i2D3
SMIL [2H]C([2H])([2H])OC(=O)Nc1nc2cc(SCCC)ccc2[nH]1
IUPAC navn trideuteriomethyl N-(5-propylsulfanyl-1H-benzimidazol-2-yl)carbamate
Molekylvægt (g/mol) 268.35
Formel vægt 268.1073