Få mere at vide
Albendazole-2-aminosulfone D3 hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30326824
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30326824 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326824 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA10065221AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Albendazole-2-amino-sulphone-D3 Hydrochloride
CAS 1435902-07-0
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C10 D3 H10 N3 O2 S . Cl H
Mængde 1 mL
Synonym Albendazole-2-aminosulfone D3 (Propyl-3,3,3 D3) Hydrochloride, 5-(3,3,3-Trideuteriopropylsulfonyl)-1H-benzimidazol-2-amine hydrochloride, Albendazole-2-aminosulfone D3 hydrochloride
InChI formel InChI=1S/C10H13N3O2S.ClH/c1-2-5-16(14,15)7-3-4-8-9(6-7)13-10(11)12-8;/h3-4,6H,2,5H2,1H3,(H3,11,12,13);1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])CCS(=O)(=O)c1ccc2[nH]c(N)nc2c1
IUPAC navn 5-(3,3,3-trideuteriopropylsulfonyl)-1H-benzimidazol-2-amine;hydrochloride
Molekylvægt (g/mol) 278.774
Formel vægt 278.068 g/mol