Få mere at vide
Albaconazole-d3, TRC
Artikelnummer. 30324225
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30324225 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324225 Leverandør Toronto Research Chemicals Leverandørnr. TRCA51145210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Albaconazole-d3
CAS 1217825-34-7
Molekylær formel C20H13D3ClF2N5O2
Mængde 10 mg
Synonym 7-Chloro-3-[(1R,2R)-2-(2,4-difluorophenyl)-2-hydroxy-1-(methyl-d3)-3-(1H-1,2,4-triazol-1-yl)propyl]-4(3H)-quinazolinone, UR 9825-d3
InChI formel InChI=1S/C20H16ClF2N5O2/c1-12(28-11-25-18-6-13(21)2-4-15(18)19(28)29)20(30,8-27-10-24-9-26-27)16-5-3-14(22)7-17(16)23/h2-7,9-12,30H,8H2,1H3/t12-,20-/m1/s1/i1D3
SMIL [2H]C([2H])([2H])[C@@H](N1C=Nc2cc(Cl)ccc2C1=O)[C@](O)(Cn3cncn3)c4ccc(F)cc4F
IUPAC navn 7-chloro-3-[(2R,3R)-1,1,1-trideuterio-3-(2,4-difluorophenyl)-3-hydroxy-4-(1,2,4-triazol-1-yl)butan-2-yl]quinazolin-4-one
Molekylvægt (g/mol) 434.84