missing translation for 'onlineSavingsMsg'
Learn More

β-Alanine, ≥98%, Ultrapure, Thermo Scientific™

Catalog Number 15855248
Click to view available options
Quantity:
200 g
1 kg
5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
107-95-9
C3H7NO2
89.09
MFCD00008200
UCMIRNVEIXFBKS-UHFFFAOYSA-N
beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
239
CHEBI:16958
3-aminopropanoic acid
NCCC(O)=O
This item is not returnable. View return policy
107-95-9
C3H7NO2
89.09
MFCD00008200
UCMIRNVEIXFBKS-UHFFFAOYSA-N
beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
239
CHEBI:16958
3-aminopropanoic acid
NCCC(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∽205°C (decomposition)
Linear Formula NH2CH2CH2COOH
Beilstein 906793
Packaging Plastic Bottle
Formula Weight 89.09
Percent Purity ≥98%
Density 1.437
Quantity 1 kg
Specific Gravity 1.437
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'