Få mere at vide
Al-23 Tube, Both Ends Open, O.D.: 40mm, I.D.: 34mm, Thermo Scientific™
Artikelnummer. 11384017
Mængde:
100 mm
200 mm
300 mm
600 mm
800 mm
1000 mm
Pakningsstørrelse:
1000mm
100mm
200mm
300mm
600mm
800mm
Artikelnummer. 11384017
Brand: Thermo Scientific Alfa Aesar 031577.DR
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Al-23 Tube
Navn note Both ends open
Mængde 200 mm