Få mere at vide
Al-23 Solid Round Rod, Dia.: 0.8mm, Thermo Scientific™
Artikelnummer. 11373038
Mængde:
100 mm
200 mm
250 mm
300 mm
Pakningsstørrelse:
100mm
200mm
250mm
300mm
Artikelnummer. 11373038
Brand: Thermo Scientific Alfa Aesar 032922.DW
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Al-23 Solid Round Rod
Mængde 300 mm