Få mere at vide
Al-23 Protective Tube, One End Closed, O.D.: 8mm, I.D.: 5mm, Wall.: 1.5mm, Thermo Scientific™
Mængde:
200 mm
270 mm
375 mm
530 mm
740 mm
1030 mm
1430 mm
2030 mm
Pakningsstørrelse:
1030mm
1430mm
200mm
2030mm
270mm
375mm
530mm
740mm
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Al-23 Protective Tube |
| Navn note | One end closed |
| Mængde | 1430 mm |