Få mere at vide
Al-23 Protective Tube, One End Closed, O.D.: 20mm, I.D.: 15mm, Wall.: 2.5mm, Thermo Scientific™
Artikelnummer. 11392698
Mængde:
200 mm
270 mm
375 mm
530 mm
740 mm
1030 mm
1430 mm
2030 mm
Pakningsstørrelse:
1030mm
1430mm
200mm
2030mm
270mm
375mm
530mm
740mm
Artikelnummer. 11392698
Brand: Thermo Scientific Alfa Aesar 030052.EE
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Al-23 Protective Tube
Navn note One end closed
Mængde 530 mm