Få mere at vide
Al-23 Multibore Round Tube, O.D.: 4mm, Four 0.8mm Bores, Thermo Scientific™
Mængde:
100 mm
200 mm
250 mm
300 mm
600 mm
800 mm
1000 mm
Pakningsstørrelse:
1000mm
100mm
200mm
250mm
300mm
600mm
800mm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Kemisk navn eller materiale | Al-23 Multibore Round Tube |
| Navn note | Four bores |
| Mængde | 1000 mm |