Få mere at vide
Al-23 Multibore Round Tube, O.D.: 2.8mm, Four 0.7mm Bores, Thermo Scientific™
Mængde:
100 mm
200 mm
250 mm
300 mm
Pakningsstørrelse:
100mm
200mm
250mm
300mm
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Al-23 Multibore Round Tube |
| Navn note | Four bores |
| Mængde | 100 mm |