Få mere at vide
Al-23 Multibore Round Tube, O.D.: 2.5mm, Two 0.7mm Bores, Thermo Scientific™
Artikelnummer. 11338487
Mængde:
100 mm
200 mm
250 mm
300 mm
Pakningsstørrelse:
100mm
200mm
250mm
300mm
Artikelnummer. 11338487
Brand: Thermo Scientific Alfa Aesar 032568.DR
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Al-23 Multibore Round Tube
Navn note Two bores
Mængde 200 mm