Få mere at vide
Al-23 Multibore Insulating Tube, O. D.: 6mm, Two 1.5mm Bores, Thermo Scientific™
Artikelnummer. 11368437
Mængde:
205 mm
275 mm
380 mm
560 mm
770 mm
1060 mm
1460 mm
2060 mm
Pakningsstørrelse:
1060mm
1460mm
205mm
2060mm
275mm
380mm
560mm
770mm
Artikelnummer. 11368437
Brand: Thermo Scientific Alfa Aesar 032556.ER
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Al-23 Multibore Insulating Tube
Navn note Two bores
Mængde 1060 mm