Få mere at vide
Al-23 Multibore Insulating Tube, O. D.: 6mm, Two 1.5mm Bores, Thermo Scientific™
Mængde:
205 mm
275 mm
380 mm
560 mm
770 mm
1060 mm
1460 mm
2060 mm
Pakningsstørrelse:
1060mm
1460mm
205mm
2060mm
275mm
380mm
560mm
770mm
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Al-23 Multibore Insulating Tube |
| Navn note | Two bores |
| Mængde | 560 mm |