Få mere at vide
InSphero Akura™ 384 Spheroid Microplate
Beskrivelse
- The Akura™ 384 Spheroid Microplate is our ultimate spheroid screening plate unifying robust liquid and spheroid handling with automated high-throughput screening applications.
- The flat bottom and black walls of the InSphero 384-well plate facilitate fluorescent imaging, while the special well design allows medium exchange, dosing, and endpoint determination in the same plate saving time and resources.
- All-in-one plate allowing spheroid formation, long-term culturing and analysis in one single plate
- Spheroid/organoid morphology and functionality are preserved in long-term cell culture
- Superior fluorescent imaging quality with a continuous, flat-bottom, 125 μm Polystyrene membrane, and a black-walled body
Tekniske data
Tekniske data
| Stregkode | No Barcode |
| Format | SBS Standard |
| membran | Polystyrene |
| Array | 24 x 16 |
| Farve | Black, Clear Bottom |
| Engangs | Single-use |
| Produkttype | Screening Plate |
| Materiale | Polystyrene |
| Antal Wells | 384 |
| Sterilitet | Sterile |
| Vis mere |