Få mere at vide
Invitrogen™ AKT Pathway (AKT/GSK/p70 S6K) Activation Profile Multispecies InstantOne™ ELISA Kit
Descripción
Includes
- User Manual
- InstantOne™ Assay Plate - 96 well (sealed)
- Cell Lysis Buffer (5X)
- Enhancer Solution
- Wash Buffer (10X)
- phospho-GSK3b (Ser9) Capture Antibody Reagent
- phospho-AKT 1/2/3 (Ser473) Capture Antibody Reagent
- phospho-p70S6K (Thr389) Capture Antibody Reagent
- phospho-GSK3b (Ser9) Detection Antibody Reagent
- phospho-AKT 1/2/3 (Ser473) Detection Antibody Reagent
- phospho-p70S6K (Thr389) Detection Antibody Reagent
- Positive Control Cell Lysate (lyophilized)
- Detection Reagent (TMB Substrate)
- Stop Solution
- Plate Seal
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated AKT 1/2/3 (Ser473) (human, mouse, hamster, rat), phosphorylated GSK3beta (Ser9) (human, mouse, rat) and phosphorylated p70S6K (Thr389) (human, mouse) in cell lysates. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Especificaciones
Especificaciones
| Adgangsnummer | P23443,P31749,P31751,P49841,Q9Y243 |
| Konjugeret | Biotin |
| Produkttype | ELISA Kit |
| Prøvetype | Cell Lysates |
| Målarter | Human |
| Til brug med (applikation) | ELISA |
| Til brug med (udstyr) | Colorimetric Microplate Reader |
| Gen-id (Entrez) | 10000, 207, 208, 2932, 6198 |
| Gen symbol | AKT,CWS6,PKB,PKB-ALPHA,PRKBA,RAC,RAC-ALPHA,PS6K,S6K,S6K-beta-1,S6K1,STK14A,p70 S6KA,p70(S6K)-alpha,p70-S6K,p70-alpha,GSK3B |
| Sættets indhold | Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal |
| Mostrar más |
For Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.