Få mere at vide
Invitrogen™ AKT Pathway (AKT/GSK/p70 S6K) Activation Profile Multispecies InstantOne™ ELISA Kit
Artikelnummer. 15500787
Mængde:
96 Tests
missing translation for 'unitSize'
96 tests
Este artículo no se puede devolver. Vea la política de devoluciones
This 3 target InstantOne ELISAs contain reagents for a total of 96 wells (32 wells for each target). Please contact Technical Service for a lot specific COA.
Código de producto. 15500787
missing translation for 'mfr': Invitrogen™ 858604811
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Este artículo no se puede devolver. Vea la política de devoluciones
This 3 target InstantOne ELISAs contain reagents for a total of 96 wells (32 wells for each target). Please contact Technical Service for a lot specific COA.
Includes
  • User Manual
  • InstantOne™ Assay Plate - 96 well (sealed)
  • Cell Lysis Buffer (5X)
  • Enhancer Solution
  • Wash Buffer (10X)
  • phospho-GSK3b (Ser9) Capture Antibody Reagent
  • phospho-AKT 1/2/3 (Ser473) Capture Antibody Reagent
  • phospho-p70S6K (Thr389) Capture Antibody Reagent
  • phospho-GSK3b (Ser9) Detection Antibody Reagent
  • phospho-AKT 1/2/3 (Ser473) Detection Antibody Reagent
  • phospho-p70S6K (Thr389) Detection Antibody Reagent
  • Positive Control Cell Lysate (lyophilized)
  • Detection Reagent (TMB Substrate)
  • Stop Solution
  • Plate Seal
InstantOne ELISA
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated AKT 1/2/3 (Ser473) (human, mouse, hamster, rat), phosphorylated GSK3beta (Ser9) (human, mouse, rat) and phosphorylated p70S6K (Thr389) (human, mouse) in cell lysates. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
AKT, which exists as multiple isoforms, is one of the principle kinases activated by phosphoinositide 3-kinase (PI3K). Activation of PI3K results in the generation of phosphatidylinositol (3,4,5)-triphosphate (PIP3). These lipid second messengers bind to the pleckstrin homology domain of AKT to promote its translocation to the plasma membrane for activation via phosphorylation at Thr308 and Ser473 by PDK1 and the mTOR TORC2 complex, respectively. Phosphorylation at both these sites is required for full activation of AKT Ser/Thr kinase activity. AKT phosphorylates over 50 known substrates, including GSK3b, p70S6K, AS160, PRAS40, TSC 1, TCS 2, Raf-1, Bad, the FOXO family of transcription factors, and PFK2. Due to its established critical role in numerous biological processes, AKT is a frequent target for oncology-related drug development. Glycogen Synthase Kinase 3 (GSK3) is a Ser/Thr kinase that exists as two isoforms, GSK3a and GSK3β. Unlike other kinases, phosphorylation at Ser9 and Ser21 for GSK3alpha and GSK3beta, respectively, leads to inactivation of this enzyme. GSK3 has been implicated in several diseases, including cancer and diabetes, through its phosphorylation of glycogen synthase. GSK3 has also been associated with neurodegenerative diseases such as Alzheimer's disease due to its phosphorylation of the neuronal Tau protein. p70 S6 Kinase (p70S6K) is a member of the ribosomal S6 kinase family of Ser/Thr kinases. P70 S6K activity is controlled by multiple signaling pathways, including the MAPK, phosphoinositide-3 kinase (PI3K), and mTOR pathways. This regulation results in the phosphorylation of multiple sites of p70 S6K, including Thr389 within the linker domain, which is required for full activation. Activated p70 S6K phosphorylates several residues on the S6 ribosomal protein, thereby leading to increased protein synthesis.
TRUSTED_SUSTAINABILITY
Especificaciones
Adgangsnummer P23443,P31749,P31751,P49841,Q9Y243
Konjugeret Biotin
Produkttype ELISA Kit
Prøvetype Cell Lysates
Målarter Human
Til brug med (applikation) ELISA
Til brug med (udstyr) Colorimetric Microplate Reader
Gen-id (Entrez) 10000, 207, 208, 2932, 6198
Gen symbol AKT,CWS6,PKB,PKB-ALPHA,PRKBA,RAC,RAC-ALPHA,PS6K,S6K,S6K-beta-1,S6K1,STK14A,p70 S6KA,p70(S6K)-alpha,p70-S6K,p70-alpha,GSK3B
Sættets indhold Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Etiket eller farvestof HRP
Mængde 96 Tests
Regulatorisk status RUO
Prøvevolumen Cell Lysate,20 μL
Opbevaringskrav 2°C to 8°C
Synonym AKT, GSK3B, p70 S6K
Mål Human
Test tid 1 hr. 20 min.
Samlet analysetid 1 hr. 10 min.
Mostrar más Mostrar menos
For Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado