missing translation for 'onlineSavingsMsg'
Få mere at vide

Ajuba Antibody, Novus Biologicals™

Artikelnummer. 18241033 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18241033 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18241033 Leverandør Novus Biologicals Leverandørnr. NB600468

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Ajuba Polyclonal specifically detects Ajuba in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Ajuba
Applikationer Western Blot, ELISA, Immunoprecipitation
Klassifikation Polyclonal
Koncentration 1.67 mg/ml
Konjugeret Unconjugated
Fortynding Western Blot 1:500-1:2500, ELISA 1:20000-1:80000, Immunoprecipitation 1:100
Formulering 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Genadgangsnr. Q96IF1, Q96IF1, Q96IF1, Q96IF1
Gene Alias Ajuba, jub, ajuba homolog (Xenopus laevis), MGC15563, protein ajuba
Gensymboler AJUBA
Værtsarter Rabbit
Immunogen This affinity purified Ajuba Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding aa 224-239 of Human Ajuba. (Uniprot: Q96IF1)
Molekylvægt af antigen 56.93 kDa
Oprensningsmetode Affinity Purified
Mængde 0.1 mg
Regulatorisk status RUO
Forskningsdisciplin Cell Cycle and Replication
Primær eller sekundær Primary
Gen-id (Entrez) 84962
Testspecificitet A BLAST analysis was used to suggest reactivity with this protein from human, rat, dog, mouse and chimpanzee based on 100% identity for the immunogen sequence. Cross reactivity with Ajuba protein homologues from other sources has not been determined.
Rekonstitution Reconstitute with 50 ul of distilled water to a final concentration of 1.0 mg/ml.
Målarter Human, Mouse, Rat, Canine, Primate
Indhold og opbevaring Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.