Learn More
Thermo Scientific™ Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
Brand: Thermo Scientific™ 598000000005
750.42 DKK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 1.10000kg
Description
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific recirculating chillers.Specifications
Air and fluid filter kit (35 micron) | |
ThermoFlex™ 900 or 1400 Recirculating Chiller | |
Air and Fluid Filter |