Learn More
Thermo Scientific™ Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
Brand:  Thermo Scientific™ 598000000007
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13224244
  • 894.25 DKK / Each
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
Description
Description
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific recirculating chillers.
Specifications
Specifications
Air and fluid filter kit (35 micron)
ThermoFlex™ 2500 Recirculating Chiller
Air and Fluid Filter