Learn More
Thermo Scientific™ Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
Brand: Thermo Scientific™ 598000000007
Additional Details : Weight : 0.01000kg
Description
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific recirculating chillers.Specifications
Air and fluid filter kit (35 micron) | |
ThermoFlex™ 2500 Recirculating Chiller | |
Air and Fluid Filter |