Få mere at vide
AICAR 3’,5’-Cyclic Phosphate, TRC
Artikelnummer. 30324610
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324610 25 mg
30321187 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30324610 Leverandør Toronto Research Chemicals Leverandørnr. TRCA44050025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale AICAR 3',5'-Cyclic Phosphate
CAS 35908-14-6
Molekylær formel C9 H13 N4 O7 P
Mængde 25 mg
InChI formel InChI=1S/C9H13N4O7P/c10-7-4(8(11)15)13(2-12-7)9-5(14)6-3(19-9)1-18-21(16,17)20-6/h2-3,5-6,9,14H,1,10H2,(H2,11,15)(H,16,17)/t3-,5-,6-,9-/m1/s1
SMIL NC(=O)c1c(N)ncn1[C@@H]2O[C@@H]3COP(=O)(O)O[C@H]3[C@H]2O
IUPAC navn 3-[(4aR,6R,7R,7aS)-2,7-dihydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-5-aminoimidazole-4-carboxamide
Molekylvægt (g/mol) 320.2
Formel vægt 320.0522