Få mere at vide
AGN 193109-d7, TRC
Artikelnummer. 30315823
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30315823 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315823 Leverandør Toronto Research Chemicals Leverandørnr. TRCA4270021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale AGN 193109 4-toluenyl-d7
CAS 1216429-25-2
Molekylær formel C28 2H7 H17 O2
Mængde 1 mg
Synonym 4-[2-[5,5-Dimethyl-8-[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)phenyl]-6H-naphthalen-2-yl]ethynyl]benzoic acid, 4-[2-[5,6-Dihydro-5,5-dimethyl-8-(4-methylphenyl)-2-naphthalenyl]ethynyl]benzoic acid D7 (4-toluenyl-D7), AGN 193109-d7, CD 3106-d7
InChI formel InChI=1S/C28H24O2/c1-19-4-11-22(12-5-19)24-16-17-28(2,3)26-15-10-21(18-25(24)26)7-6-20-8-13-23(14-9-20)27(29)30/h4-5,8-16,18H,17H2,1-3H3,(H,29,30)/i1D3,4D,5D,11D,12D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1C2=CCC(C)(C)c3ccc(cc23)C#Cc4ccc(cc4)C(=O)O)C([2H])([2H])[2H]
IUPAC navn 4-[2-[5,5-dimethyl-8-[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)phenyl]-6H-naphthalen-2-yl]ethynyl]benzoic acid
Molekylvægt (g/mol) 399.53
Formel vægt 399.2216