Få mere at vide
Tocris Bioscience™ AGN 193109
Beskrivelse
TB5758-RMU is an Ancillary Material Grade version of product AGN 193109 (Cat. No. 5758). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
Find out more about our Ancillary Material Grade product range.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[2-[5,6-Dihydro-5,5-dimethyl-8-(4-methylphenyl)-2-naphthalenyl]ethynyl]benzoic acid |
| CAS | 171746-21-7 |
| Mængde | 10 mg |
| Mål | Retinoic Acid Receptor Antagonists |
| Molekylær formel | C28H24O2 |
| Renhed | 0.98 |