missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ AGI 5198
GREENER_CHOICE

Artikelnummer. 18796816
Change view
Click to view available options
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18796816 25 mg
18739466 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18796816 Supplier Tocris Bioscience™ Supplier No. 7087/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective inhibitor of mutant isocitrate dehydrogenase 1 (mIDH1)

AGI 5198 is a potent and selective inhibitor of mutant isocitrate dehydrogenase 1 (mIDH1; IC50 = 70 nM). Selective for mIDH1 over wild-type IDH1 and IDH2. Inhibits production of oncometabolite D-2 hydroxyglutarate. Attenuates growth of glioma cells containing mIDH1, but not wild-type IDH1, in vitro and in vivo. Also inhibits growth of fibrosarcoma and chondrosarcoma cells bearing mIDH1.
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-[2-(Cyclohexylamino)-1-(2-methylphenyl)-2-oxoethyl]-N-(3-fluorophenyl)-2-methyl-1 H-Imidazole-1-acetamide
CAS 1355326-35-0
Mængde 25 mg
Mål Dehydrogenase Inhibitors
Molekylær formel C27H31FN4O2
Renhed 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.