Learn More
RETSCH Agate and Zirconium Oxide O-ring
Brand:  RETSCH 05.114.0070
4778.50 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11914352
  • 5030.00 DKK / Each
Estimated Shipment: 29-08-2024
to see stock.
Specifications
O-Ring
1Each
Agate and Zirconium Oxide
For Vibratory Disc Mill RS 200 100mL Grinding Set