missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Agarose, low EEO
630.97 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
100 g
Description
Agarose is useful in separation of nucleic acids electrophoretically, Suitable for isoelectric focusing, protein blotting and antibody separation Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Agarose is useful in separation of nucleic acids electrophoretically, Suitable for isoelectric focusing, protein blotting and antibody separation Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.
Solubility
Soluble in water
Notes
Gel strength: >2400g/cm² (1.5% w/v); Gelling temperature: 35-37°C; EEO: 0.07-0.1
Agarose is useful in separation of nucleic acids electrophoretically, Suitable for isoelectric focusing, protein blotting and antibody separation Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.
Solubility
Soluble in water
Notes
Gel strength: >2400g/cm² (1.5% w/v); Gelling temperature: 35-37°C; EEO: 0.07-0.1
Specifications
Physical Form | Powder |
Melting Point | 87°C to 89°C |
Color | White |
Quantity | 50 g |
Solubility Information | Soluble in water |
Sensitivity | Hygroscopic; Moisture sensitive |
Chemical Name or Material | Agarose, low EEO |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'