missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agar, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 lb.
5 lb.
Description
- Light tan granular powder
- CAS: 9002-18-0
- MDL Number: MFCD00081288
Specifications
Color | Light tan |
Form | Granulated Powder |
Product Type | Agar |
For Use With (Application) | Used as a Gelling Agent in Preparation of Culture Media. |
Packaging Type | Plastic Bottle |
Quantity | 5 lb. |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'