Learn More
Xylem AG Refractometer Calibration Fluid 20 x 5 mL (UKAS/NIST)
Calibration fluid for calibration of refractometers.
Brand: Xylem 90-602
This item is not returnable.
View return policy
Description
- Calibration Fluid
- For Refractometers
Specifications
Calibration Fluid | |
1.34026 | |
Refractometer |
Product Content Correction
Product Title
Xylem AG Refractometer Calibration Fluid 20 x 5 mL (UKAS/NIST) > 1.34026
Spot an opportunity for improvement?Share a Content Correction