Få mere at vide
Afloqualone, MedChemExpress
Artikelnummer. 16478816
Encompass_Preferred
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
16478816 100 mg
1 options
Artikelnummer. 16478816 Leverandør Medchem Express Leverandørnr. HYB1833
MedChemExpress Afloqualone (HQ-495) is a GABAergic agent and has agonist activity at the β subtype of the GABAα receptor. Afloqualone has antivertiginous effects thought to be attributable to the increased sensitivity of GABA receptors of the LVN neuron site.
Afloqualone (HQ-495) is a GABAergic agent and has agonist activity at the β subtype of the GABAα receptor. Afloqualone has antivertiginous effects thought to be attributable to the increased sensitivity of GABA receptors of the LVN neuron site.
Tekniske data
Kemisk navn eller materiale Afloqualone
CAS 56287-74-2
Farve White
Renhedsgrad noter Research
Molekylær formel C16H14FN3O
Mængde 100 mg
Synonym HQ-495
Opløselighedsinformation DMSO : ≥ 33 mg/mL (116.48 mM)
SMIL O=C1N(C2=CC=CC=C2C)C(CF)=NC3=C1C=C(N)C=C3
Molekylvægt (g/mol) 283.3
Formel vægt 283.3
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation