Få mere at vide
Aflatoxin Q1, TRC
Artikelnummer. 30318854
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30318854 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318854 Leverandør Toronto Research Chemicals Leverandørnr. TRCA3575300.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Aflatoxin Q1
CAS 52819-96-2
Molekylær formel C17 H12 O7
Mængde 0.25 mg
Synonym (3S,6aR,9aS)-2,3,6a,9a-Tetrahydro-3-hydroxy-4-methoxycyclopenta[c]furo[3',2':4,5]furo[2,3-h][1]benzopyran-1,11-dione, [3S-(3α,6aα,9aα)]-2,3,6a,9a-Tetrahydro-3-hydroxy-4-methoxycyclopenta[c]furo[3',2':4,5]furo[2,3-h][1]benzopyran-1,11-dione
InChI formel InChI=1S/C17H12O7/c1-21-9-5-10-11(6-2-3-22-17(6)23-10)15-14(9)12-7(18)4-8(19)13(12)16(20)24-15/h2-3,5-7,17-18H,4H2,1H3/t6-,7-,17+/m0/s1
SMIL COc1cc2O[C@H]3OC=C[C@H]3c2c4OC(=O)C5=C([C@@H](O)CC5=O)c14
IUPAC navn (3S,7R,14S)-14-hydroxy-11-methoxy-6,8,19-trioxapentacyclo[10.7.0.02,9.03,7.013,17]nonadeca-1,4,9,11,13(17)-pentaene-16,18-dione
Molekylvægt (g/mol) 328.27
Formel vægt 328.0583