Få mere at vide
Tocris Bioscience™ Afatinib Dimaleate
Beskrivelse
Afatinib dimaleate is a potent dual specificity EGFR/HER2 inhibitor (IC50 are 0.4, 0.5, 10, 14, 1 nM for EGFRL858R, EGFRwt, EGFR L858R/T790M, ErbB2 (HER2) and ErbB4 (HER4), respectively). 100-fold more active than Gefitinib in L858R-T790M EGFR double mutant cell line. Irreversibly binds EGFR and HER2. Suppresses the kinase activity of wild-type and activated EGFR and HER2 mutants, including erlotinib-resistant isoforms. Induces tumor regression in xenograft and transgenic lung cancer models.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[[(3S)-tetrahydro-3-furanyl]oxy]-6-quinazolinyl]-4-(dimethylamino)-2-butenamide dimaleate |
| CAS | 850140-73-7 |
| Synonym | BIBW2992 |
| Mængde | 10 mg |
| Molekylær formel | C24H25ClFN5O3.2C4H4O4 |
| Formel vægt | 718.08 |
| Renhed | 98% |