Få mere at vide
Afatinib-d6, TRC
Artikelnummer. 30309975
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30309975 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309975 Leverandør Toronto Research Chemicals Leverandørnr. TRCA3553031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Afatinib-d6
CAS 1313874-96-2
Molekylær formel C24H19D6ClFN5O3
Mængde 1 mg
Synonym (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[[(3S)-tetrahydro-3-furanyl]oxy]-6-quinazolinyl]-4-(dimethylamino)-2-butenamide-d6, BIBW 2992-d6
InChI formel InChI=1S/C24H25ClFN5O3/c1-31(2)8-3-4-23(32)30-21-11-17-20(12-22(21)34-16-7-9-33-13-16)27-14-28-24(17)29-15-5-6-19(26)18(25)10-15/h3-6,10-12,14,16H,7-9,13H2,1-2H3,(H,30,32)(H,27,28,29)/b4-3+/t16-/m0/s1/i1D3,2D3
SMIL ClC1=C(F)C=CC(NC2=NC=NC3=CC(O[C@H]4CCOC4)=C(NC(/C=C/CN(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)C=C32)=C1
IUPAC navn (E)-4-[bis(trideuteriomethyl)amino]-N-[4-(3-chloro-4-fluoroanilino)-7-[(3S)-oxolan-3-yl]oxyquinazolin-6-yl]but-2-enamide
Molekylvægt (g/mol) 491.98