missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ AF1Q Recombinant Protein

Brand:  Abnova™ H00010962-P01.10ug

 View more versions of this product

Product Code. 16135932

  • 2735.00 DKK / 10µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH09624
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
35.64
8
Glutathione Sepharose 4 Fast Flow
10 μg
MRDPVSSQYSSFLFWRMPIPELDLSELEGLGLSDTATYKVKDSSVGKMIGQATAADQEKNPEGDGLLEYSTFNFWRAPIASIHSFELDLL
AF1Q/RP11-316M1.10
MLLT11
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10962
AF1Q (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MLLT11
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ AF1Q Recombinant Protein > 10μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.