Få mere at vide
Tocris Bioscience™ AF 64394
GREENER_CHOICE
Artikelnummer. 18719966
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18719966 10 mg
18789276 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18719966 Leverandør Tocris Bioscience™ Leverandørnr. 6931/10
Potent and selective GPR3 inverse agonist
AF 64394 is a potent and selective GPR3 inverse agonist (pIC50 = 7.3). Selective for GPR3 over GPR6 and GPR12 (pIC50 values are 5.1 and 4.9, respectively).
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[[4-Chloro-2-(1-methylethoxy)phenyl]methyl]-5-phenyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
CAS 1637300-25-4
Mængde 10 mg
Molekylær formel C21H20ClN5O
Renhed 98%