missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Adrenorphin, Free acid

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66389.M¬

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15487729

  • 469.11 DKK / 1mg


Estimated Shipment
21-10-2020
Add to basket

Specifications

Specifications

985.16
1mg
Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val
-30° to -10°C
HPLC
Solid
RUO
Adrenorphin, free acid
Videos
SDS Documents

For Research Use Only