missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adrenocorticotropic Hormone (1-24), human
Click to view available options
Quantity:
0.5 mg
1 mg
Description
Adrenocorticotropic Hormone (1-24), human is used in the treatment of West syndrome. It activates G proteins. It stimulates secretion of glucocorticoid steroid hormones from adrenal cortex cells. It also enhances transcription of mitochondrial genes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Content And Storage | Store at -20°C |
Format | Solid |
Quantity | 0.5 mg |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'