missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adrenocorticotropic Hormone (1-24), human

Catalog Number p-7076668
Click to view available options
Quantity:
0.5 mg
1 mg
This item is not returnable. View return policy
16960-16-0
This item is not returnable. View return policy
16960-16-0

Used in the treatment of West syndrome. It activates G proteins. It stimulates secretion of glucocorticoid steroid hormones from adrenal cortex cells. It also enhances transcription of mitochondrial genes.

Adrenocorticotropic Hormone (1-24), human is used in the treatment of West syndrome. It activates G proteins. It stimulates secretion of glucocorticoid steroid hormones from adrenal cortex cells. It also enhances transcription of mitochondrial genes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Content And Storage Store at -20°C
Format Solid
Quantity 0.5 mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'