missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK Antibody (001), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90184H
Additional Details : Weight : 0.00970kg
Description
ADK Monoclonal specifically detects ADK in Human samples. It is validated for ELISA.Specifications
ADK | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1 | |
ELISA | |
adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ADK (AAH03568.1; Met1-His345). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism | |
132 | |
Store at 4°C in the dark. | |
IgG |